Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AICDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049116.1 | XP_017904605.1 | 1 - 2 | 149 | 61 | |
GenBank mRNA | JT642702.1 | - | 2 - 3 | 159 | 61 | |
JT689631.1 | - | 2 - 3 | 223 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |