Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050110.1 | XP_017905599.1 | 4 - 5 | 755 | 62 | |
XM_018050111.1 | XP_017905600.1 | 3 - 4 | 700 | 62 | ||
GenBank mRNA | KA331293.1 | - | 4 - 5 | 799 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |