Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018067054.1 | XP_017922543.1 | 3 - 4 | 233 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |