Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102180151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064520.1 | XP_017920009.1 | 3 - 4 | 1496 | 109 | |
XM_018064522.1 | XP_017920011.1 | 3 - 4 | 994 | 109 | ||
GenBank mRNA | KA312316.1 | - | 1 - 2 | 580 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Capra hircus Probe spans exons |