Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040868.1 | XP_017896357.1 | 3 - 4 | 789 | 73 | |
XM_018040876.1 | XP_017896365.1 | 3 - 4 | 440 | 73 | ||
XM_018040883.1 | XP_017896372.1 | 2 - 3 | 308 | 73 | ||
XM_018040891.1 | XP_017896380.1 | 2 - 3 | 280 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |