Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057646.1 | XP_017913135.1 | 6 - 7 | 1209 | 70 | |
XM_018057647.1 | XP_017913136.1 | 5 - 6 | 1138 | 70 | ||
XM_018057649.1 | XP_017913138.1 | 5 - 6 | 584 | 70 | ||
XM_018057650.1 | XP_017913139.1 | 6 - 7 | 650 | 70 | ||
XM_018057651.1 | XP_017913140.1 | 3 - 4 | 347 | 70 | ||
GenBank mRNA | JO645466.2 | - | 5 - 6 | 614 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |