Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VOPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038387.1 | XP_017893876.1 | 4 - 5 | 457 | 65 | |
XM_018038388.1 | XP_017893877.1 | 3 - 4 | 381 | 65 | ||
GenBank mRNA | JO448084.2 | - | 4 - 5 | 456 | 65 | |
JO593067.2 | - | 4 - 5 | 467 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |