Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040962.1 | XP_017896451.1 | 4 - 5 | 532 | 114 | |
GenBank mRNA | JO375465.2 | - | 1 - 2 | 161 | 114 | |
JO412308.2 | - | 4 - 5 | 538 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |