Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685128.3 | XP_005685185.1 | 1 - 2 | 162 | 91 | |
XM_013966771.2 | XP_013822225.1 | 1 - 2 | 162 | 91 | ||
XM_018053618.1 | XP_017909107.1 | 1 - 2 | 162 | 91 | ||
XM_018053619.1 | XP_017909108.1 | 1 - 2 | 162 | 91 | ||
XM_018053620.1 | XP_017909109.1 | 1 - 2 | 160 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |