Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045858.1 | XP_017901347.1 | 6 - 7 | 574 | 79 | |
GenBank mRNA | JO388976.2 | - | 3 - 4 | 411 | 79 | |
JO579974.1 | - | 3 - 4 | 471 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |