Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680853.3 | XP_005680910.2 | 2 - 3 | 1275 | 98 | |
GenBank mRNA | JT762615.1 | - | 4 - 5 | 827 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |