Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053892.1 | XP_017909381.1 | 3 - 4 | 756 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |