Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHACTR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057602.1 | XP_017913091.1 | 3 - 4 | 807 | 54 | |
XM_018057603.1 | XP_017913092.1 | 3 - 4 | 351 | 54 | ||
GenBank mRNA | JO622554.2 | - | 10 - 11 | 1726 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |