Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693302.3 | XP_005693359.1 | 3 - 4 | 746 | 98 | |
GenBank mRNA | KA323109.1 | - | 3 - 4 | 738 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |