Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXLNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043859.1 | XP_017899348.1 | 1240 | 108 | ||
GenBank mRNA | JO593009.2 | - | 1166 | 108 | ||
JO593010.2 | - | 1166 | 108 |
Gene Symbol: LOC102175567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045074.1 | XP_017900563.1 | 1 - 2 | 690 | 108 | |
GenBank mRNA | JO642386.2 | - | 717 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |