Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044816.1 | XP_017900305.1 | 2 - 3 | 164 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |