Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063699.1 | XP_017919188.1 | 8 - 9 | 1665 | 64 | |
XM_018063700.1 | XP_017919189.1 | 8 - 9 | 1693 | 64 | ||
XM_018063701.1 | XP_017919190.1 | 8 - 9 | 1617 | 64 | ||
XM_018063702.1 | XP_017919191.1 | 8 - 9 | 1668 | 64 | ||
XM_018063703.1 | XP_017919192.1 | 8 - 9 | 1671 | 64 | ||
XM_018063704.1 | XP_017919193.1 | 8 - 9 | 1674 | 64 | ||
GenBank mRNA | JO577302.1 | - | 8 - 9 | 1524 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |