Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHNAK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065939.1 | XP_017921428.1 | 7 | 9128 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |