Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DISP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690504.3 | XP_005690561.2 | 4 - 5 | 880 | 64 | |
XM_013970119.2 | XP_013825573.2 | 4 - 5 | 765 | 64 | ||
XM_018060190.1 | XP_017915679.1 | 4 - 5 | 792 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |