Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061435.1 | XP_017916924.1 | 5 - 6 | 1668 | 89 | |
XM_018061436.1 | XP_017916925.1 | 5 - 6 | 1253 | 89 | ||
XM_018061437.1 | XP_017916926.1 | 5 - 6 | 1291 | 89 | ||
GenBank mRNA | JO423385.2 | - | 1 - 2 | 260 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |