Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692819.3 | XP_005692876.2 | 2 - 3 | 466 | 71 | |
XM_013971511.2 | XP_013826965.1 | 2 - 3 | 431 | 71 | ||
XM_013971512.2 | XP_013826966.1 | 2 - 3 | 150 | 71 | ||
GenBank mRNA | JO581400.1 | - | 2 - 3 | 552 | 71 | |
JO581401.1 | - | 2 - 3 | 549 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |