Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041766.1 | XP_017897255.1 | 2 - 3 | 7373 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |