Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066531.1 | XP_017922020.1 | 8 - 9 | 1435 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |