Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDXACB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689433.3 | XP_005689490.2 | 1 - 2 | 206 | 100 | |
GenBank mRNA | KA311210.1 | - | 1 - 2 | 127 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |