Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063939.1 | XP_017919428.1 | 5 - 6 | 1450 | 92 | |
XM_018063940.1 | XP_017919429.1 | 6 - 7 | 1578 | 92 | ||
XM_018063941.1 | XP_017919430.1 | 5 - 6 | 1633 | 92 | ||
GenBank mRNA | JO427631.1 | - | 6 - 7 | 1676 | 92 | |
JO428131.1 | - | 5 - 6 | 1547 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |