Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFHR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691048.3 | XP_005691105.2 | 2 - 3 | 342 | 78 | |
XM_013970400.2 | XP_013825854.2 | 2 - 3 | 342 | 78 | ||
GenBank mRNA | JO412079.2 | - | 2 - 3 | 369 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |