Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042481.1 | XP_017897970.1 | 11 - 12 | 5603 | 70 | |
XM_018042482.1 | XP_017897971.1 | 11 - 12 | 5603 | 70 | ||
XM_018042483.1 | XP_017897972.1 | 10 - 11 | 5516 | 70 | ||
GenBank mRNA | KU870424.1 | - | 10 - 11 | 5253 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |