Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064594.1 | XP_017920083.1 | 1 - 2 | 627 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |