Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC48A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047881.1 | XP_017903370.1 | 2 - 3 | 430 | 66 | |
GenBank mRNA | JO556425.2 | - | 2 - 3 | 420 | 66 | |
JO632180.2 | - | 1 - 2 | 2327 | 66 | ||
KA340085.1 | - | 1 - 2 | 2165 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |