Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEDD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013966838.1 | XP_013822292.1 | 4 - 5 | 478 | 58 | |
GenBank mRNA | JO579493.1 | - | 1 - 2 | 437 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |