Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTHRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013969243.2 | XP_013824697.1 | 2 - 3 | 547 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |