Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041456.1 | XP_017896945.1 | 13 - 14 | 2745 | 71 | |
XM_018041457.1 | XP_017896946.1 | 13 - 14 | 2745 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |