Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678109.3 | XP_005678166.3 | 5 - 6 | 1130 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |