Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16H1orf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060709.1 | XP_017916198.1 | 2 - 3 | 361 | 70 | |
XM_018060710.1 | XP_017916199.1 | 2 - 3 | 581 | 70 | ||
GenBank mRNA | JO392940.2 | - | 2 - 3 | 513 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |