Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063327.1 | XP_017918816.1 | 3 - 4 | 4955 | 65 | |
XM_018063328.1 | XP_017918817.1 | 3 - 4 | 4955 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Capra hircus Probe spans exons |