Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046711.1 | XP_017902200.1 | 1 - 2 | 480 | 70 | |
XM_018046714.1 | XP_017902203.1 | 1 - 2 | 483 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |