Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693239.3 | XP_005693296.1 | 4 - 5 | 919 | 84 | |
GenBank mRNA | JO410291.2 | - | 4 - 5 | 896 | 84 | |
JO410292.2 | - | 4 - 5 | 896 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |