Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064790.1 | XP_017920279.1 | 4 - 5 | 2178 | 86 | |
XM_018064800.1 | XP_017920289.1 | 4 - 5 | 1603 | 86 | ||
XM_018064805.1 | XP_017920294.1 | 3 - 4 | 1591 | 86 | ||
GenBank mRNA | JO339810.2 | - | 3 - 4 | 1317 | 86 | |
JO428667.2 | - | 3 - 4 | 901 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |