Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACTBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018067081.1 | XP_017922570.1 | 2 - 3 | 257 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |