Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2H2orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676445.2 | XP_005676502.2 | 7 - 8 | 656 | 107 | |
GenBank mRNA | JT671864.1 | - | 1 - 2 | 385 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |