Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691856.3 | XP_005691913.1 | 2 - 3 | 439 | 94 | |
GenBank mRNA | JO402918.2 | - | 6 - 7 | 681 | 94 | |
JO402919.2 | - | 5 - 6 | 693 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |