Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005700732.3 | XP_005700789.1 | 3 - 4 | 930 | 76 | |
XM_013976472.2 | XP_013831926.1 | 2 - 3 | 569 | 76 | ||
XM_013976477.2 | XP_013831931.1 | 3 - 4 | 253 | 76 | ||
XM_013976478.2 | XP_013831932.1 | 3 - 4 | 227 | 76 | ||
XM_018043992.1 | XP_017899481.1 | 5 - 6 | 2123 | 76 | ||
XM_018043993.1 | XP_017899482.1 | 5 - 6 | 2156 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |