Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013972164.2 | XP_013827618.1 | 1 - 2 | 261 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |