Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689440.3 | XP_005689497.2 | 5 - 6 | 494 | 131 | |
XM_005689441.3 | XP_005689498.2 | 5 - 6 | 453 | 131 | ||
XM_005689442.3 | XP_005689499.2 | 6 - 7 | 464 | 131 | ||
GenBank mRNA | JO424465.1 | - | 1 - 2 | 1707 | 131 | |
JT707918.1 | - | 7 - 8 | 842 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |