Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANGRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693555.3 | XP_005693612.1 | 2 - 3 | 341 | 90 | |
XM_005693556.3 | XP_005693613.1 | 2 - 3 | 345 | 90 | ||
GenBank mRNA | JO373137.2 | - | 2 - 3 | 342 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |