Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAVER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013962779.2 | XP_013818233.2 | 4 - 5 | 1338 | 74 | |
XM_013962780.2 | XP_013818234.2 | 4 - 5 | 1339 | 74 | ||
XM_013962781.2 | XP_013818235.2 | 4 - 5 | 1340 | 74 | ||
XM_018045157.1 | XP_017900646.1 | 4 - 5 | 1339 | 74 | ||
XM_018045163.1 | XP_017900652.1 | 4 - 5 | 1342 | 74 | ||
GenBank mRNA | JT646385.1 | - | 9 - 10 | 1419 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |