Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043072.1 | XP_017898561.1 | 9 - 10 | 1960 | 66 | |
GenBank mRNA | KA344435.1 | - | 255 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |