Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684627.3 | XP_005684684.2 | 4 - 5 | 1858 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |