Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKZF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698578.3 | XP_005698635.1 | 2 - 3 | 342 | 70 | |
XM_005698579.3 | XP_005698636.1 | 3 - 4 | 464 | 70 | ||
XM_018041367.1 | XP_017896856.1 | 3 - 4 | 591 | 70 | ||
GenBank mRNA | JO604437.2 | - | 1 - 2 | 152 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |