Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018067086.1 | XP_017922575.1 | 3 - 4 | 1337 | 70 | |
GenBank mRNA | JT673937.1 | - | 3 - 4 | 1012 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |